Bægre, lav eller høj type, borosilikatglas
Leverandør: Brand
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
90617 90624 90629 90636 90641 90648 90654 90663 91210 91212 91214 91217 91224 91236 91241 91248 91253 91254 91263 91268 91273 91229 90668
BRND90617EA 999 DKK
BRND90617 BRND90624 BRND90629 BRND90636 BRND90641 BRND90648 BRND90654 BRND90663 BRND91210 BRND91212 BRND91214 BRND91217 BRND91224 BRND91236 BRND91241 BRND91248 BRND91253 BRND91254 BRND91263 BRND91268 BRND91273 BRND91229 BRND90668
Bægre, lav eller høj type, borosilikatglas
Bægre
Borosilicate glass 3.3.
  • Very good chemical resistance
  • High temperature resistance
  • Autoclavable
  • Beakers 5 ml and 10 ml without graduations and labeling area
Volume graduations and labelling area.
Certificeringer: DIN 12331, ISO 3819