Acetonitril-D3 (99.8% D)
Leverandør: CAMBRIDGE ISOTOPE LABS
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
DLM-21-10
DLM-21-10X0.75
CAMBDLM-21-10EA
0
DKK
CAMBDLM-21-10
CAMBDLM-21-10X0.75
Acetonitril-D3 (99.8% D)
Acetonitril-D3
Formel:
D₃CCN Kogepunkt: 80,7 °C (1013 hPa) Smeltepunkt: –42 °C Massefylde: 0,844 g/cm³ (20 °C) Flammepunkt: 2 °C (closed cup) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00001881 CAS-nummer: 2206-26-0 EINECS: 218-616-5 UN nummer: 1648 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...