2-Methylcyclohexanol cis- and trans-mixture ≥97%
Leverandør: Thermo Fisher ScientificSamlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Purity: 97%
CAS No.: 583-59-5
Molecular Formula: C7H14O
Formula Weight: 114.19
Flash Point: 58°(136°F)
Refractive Index: 1.4620
Melting Point: -21°
Boiling Point: 163-166°
Beilstein Reference: 1847535
UN No.: 2617
MDL No.: MFCD00001443
Formel:
C₇H₁₄O Kogepunkt: 163…166 °C Smeltepunkt: –21 °C Massefylde: 0,929 Flammepunkt: 58 °C (136 °F) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00001443 CAS-nummer: 583-59-5 EINECS: 209-512-0 UN nummer: 2617 ADR: 3,III |