Centrifuge tube filters, Costar® Spin-X®
Leverandør: Corning
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
8160 8161 8162 8163 8169 8170 9301
525-0017EA 2570 DKK
525-0017 525-0018 525-0019 525-0020 525-3415 525-3416 612-5201
Centrifuge tube filters, Costar® Spin-X®
Filtre Centrifugefiltre
Spin-X® centrifuge tube filters use centrifugation to remove bacteria, particles, or cells from liquids, HPLC sample preparation and DNA removal from agarose or acrylamide gels.
  • Cellulose acetate (CA) membranes provide fast flow rates and low protein binding
  • Sterile and nonpyrogenic and DNase-/RNase-free
  • Maximum relative centrifugal force is 16000×g
Units consist of a membrane-containing insert and a polypropylene centrifuge tube. Choose the cellulose acetate membrane insert for aqueous solutions or the nylon membrane insert for HPLC applications.More Product Information