Nalgene® Diagnostics Bottles, Narrow Neck, Round, HDPE, Amber, with Screw Caps
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
312004-9050 312084-9025 312004-9025 312004-9125 312084-9050 312084-9125
NALG312004-9050EA 24100 DKK
NALG312004-9050 NALG312084-9025 NALG312004-9025 NALG312004-9125 NALG312084-9050 NALG312084-9125
Nalgene® Diagnostics Bottles, Narrow Neck, Round, HDPE, Amber, with Screw Caps
Flasker Plastflasker
Bottles with closure, suitable for storage and shipping of small volume, light sensitive and valuable reagents.
  • Non-cytotoxic and leakproof
  • Opaque, linerless polypropylene closures
  • Durable, uniform walls that resist splitting and puncturing
Bottles provide light resistance with the ability to see liquid level and are ideal for test kits and many small-volume applications.
Certificeringer: Compliance with ISO 13485:2003; Meets the requirement of 21CFR177.1520 and USP Class VI.
Bestillingsoplysninger: Product certificate providing cytotoxicity,More Product Information