Skilletragte
Leverandør: Brand
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
140004 140005 140006 140007
BRND140004EA 1840 DKK
BRND140004 BRND140005 BRND140006 BRND140007
Skilletragte
Tragte Skilletragte
PP, high clarity, PTFE stopcock, PP screw cap.
  • Leakproof closure and stopcock
  • Excellent chemical resistance
  • Autoclavable except for stopcock assembly
  • PTFE stopcock requires no lubrication
High clarity for observation of contents.