Flasker, smal hals, Boston type, HDPE, med skruelåg, Nalgene®
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
312004-0016 312004-0004 312004-0001 312004-0002 312004-0032 312004-0008
215-2559EA 4810 DKK
215-2559 215-2557 215-2555 215-2556 215-2560 215-2558
Flasker, smal hals, Boston type, HDPE, med skruelåg, Nalgene®
Flasker Plastflasker
HDPE, amber with amber PP screw cap.
  • Good chemical resistance
  • Heat resistant up to +120 °C
  • Leakproof
Amber to protect light-sensitive materials. Ideal for storing and shipping liquids.