PCR plates for Roche® LightCycler® 480
Leverandør: Brand
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
781358 781365 781364 781358BC
731-0164EA 4190 DKK
731-0164 732-1463 732-1462 731-0439
PCR plates for Roche® LightCycler® 480
Mikroplader PCR plader
PP, white. Available with 96-well low profile and 384-well formats. Both plates are compatible with many thermal cyclers, but are perfectly suited for use in the Roche® LightCycler® 480.
  • Uniformally coloured with titanium dioxide (TiO₂)
  • Smooth surfaces to optimise reflection of the fluorescence signal
  • Extra-thin wall thickness to provide optimal thermal transfer and short cycle times
  • 96-well plates have black alphanumeric coding, no additional markings on the cut corner
  • Free from DNase, RNase and DNA
  • Autoclavable 121 °C (2 bar), acc. DIN EN 285
*suitable for multichannel pipettes and robotic systems.
Bestillingsoplysninger: Barcoded versions of these plates are also available. Please contact your local Avantor sales office for further details.