Chrom ≥99% (metals basis), powder -100 mesh
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Advarsel
10147.A3 10147.09 10147.22 10147.36
10147.A3EA 14920 DKK
10147.A3 10147.09 10147.22 10147.36
Chrom ≥99% (metals basis), powder -100 mesh
Chromium
Purity: 99% CAS No.: 7440-47-3 Physical Form: Apparent density 2.0-3.0g/cm3 Appearance: Silver Odour: Odourless UN No.: 3089 MDL No.: MFCD00010944
Formel: Cr
Molekylvægt: 52 g/mol
Kogepunkt: 2469
Smeltepunkt: 1840
Massefylde: 7.2
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00010944
CAS-nummer: 7440-47-3
EINECS: 231-157-5
UN nummer: 3089
ADR: 4.1 III
Merck Index: 12,02288