Zink ≥99.9% (metals basis), powder -100 mesh

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

00424.A1 00424.A7 00424.30
00424.A1EA 1580 DKK
00424.A1 00424.A7 00424.30
Zink ≥99.9% (metals basis), powder -100 mesh
Zinc

Purity: 99.9% CAS No.: 7440-66-6 Sensitivity: Air and Moisture Sensitive Appearance: Grey Odour: Odourless UN No.: 1436 MDL No.: MFCD00011291

Formel: Zn
Kogepunkt: 907 °C (1013 hPa)
Massefylde: 7,13 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00011291
CAS-nummer: 7440-66-6
EINECS: 231-175-3
UN nummer: 1436
ADR: 4.3,III

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR