Dunke, med sanitetsflanger, Nalgene®
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
2640-0020 2640-0050 2640-0130
NALG2640-0020EA 6120 DKK
NALG2640-0020 NALG2640-0050 NALG2640-0130
Dunke, med sanitetsflanger, Nalgene®
Flasker Flasker, dunke
Made of PP, translucent.
  • Graduated to contain in liters and gallons
  • With thermoplastic elastomer gasket
  • Molded-in handles
  • Excellent chemical and temperature resistance
Use the autoclavable PP Carboys with 1,5" sanitary flanges as dispensing ports or with secure hookup for supply reservoirs to systems such as fermentors or chromatography columns.