Magnetiske omrørerstave, PTFE, ottekantede, med midterring
Leverandør: Brand
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
137607 137608 137610 137612 137615 137617 137618 137620 137622 137625
BRND137607EA 531 DKK
BRND137607 BRND137608 BRND137610 BRND137612 BRND137615 BRND137617 BRND137618 BRND137620 BRND137622 BRND137625
Magnetiske omrørerstave, PTFE, ottekantede, med midterring
Omrørere Omrøremagneter
These octagonal shaped PTFE magnetic stirrer bars are ideal for vessels with slightly uneven bottoms. The angular surface creates high turbulence even at low speeds.
Designed with pivot ring structure which reduces the contact surface and enables the stirrer to adopt the optimum position.