Sikkerhedskander, LDPE, Nalgene®
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
2340-0020 2340-0050
215-1124EA 2510 DKK
215-1124 119-3125
Sikkerhedskander, LDPE, Nalgene®
Flasker Flasker, dunke
LDPE, translucent, with PP screw cap.
  • Pour spout reduces splash; back off closure to vent
  • Spout plug permanentelly attached to jug
  • Leakproof
Wide shoulder handle for easy carrying. Integral, moulded bottom grip gives additional support when pouring.