Magnetic stirring bars, PTFE, star-shaped, Spinstar®
Leverandør: Brand
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
137810 137812 137815 137817 137820 137822 137826
BRND137810EA 564 DKK
BRND137810 BRND137812 BRND137815 BRND137817 BRND137820 BRND137822 BRND137826
Magnetic stirring bars, PTFE, star-shaped, Spinstar®
Omrørere Omrøremagneter
Triangular shaped PTFE magnetic stirring bars.
  • Ideal for dissolving solids and mixing sediments due to scraping action
  • Angular surface creates high turbulence even at low speeds