Zink ≥99.95% (metals basis), wire, Ø 2.0 mm (0.08 in)

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

42706.G1 42706.G5 42706.G9
42706.G1EA 684 DKK
42706.G1 42706.G5 42706.G9
Zink ≥99.95% (metals basis), wire, Ø 2.0 mm (0.08 in)
Zinc

Purity: 99.95% CAS No.: 7440-66-6 Physical Form: ≈22.4g/m Odour: Odourless MDL No.: MFCD00011291

Formel: Zn
Kogepunkt: 908 °C (1013 hPa)
Smeltepunkt: 420 °C
Massefylde: 7,14 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00011291
CAS-nummer: 7440-66-6
EINECS: 231-175-3
Merck Index: 12,10255

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR