Zink ≥99.9985% (metals basis), wire, Puratronic®, Ø 1.0 mm (0.04 in)
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
12053.BW
12053.CB
12053.BWEA
782
DKK
12053.BW
12053.CB
Zink ≥99.9985% (metals basis), wire, Puratronic®, Ø 1.0 mm (0.04 in)
Zinc
Purity: 99.9985%
CAS No.: 7440-66-6
Physical Form: ≈5.60g/m
Odour: Odourless
MDL No.: MFCD00011291
Formel:
Zn Kogepunkt: 908 °C (1013 hPa) Smeltepunkt: 420 °C Massefylde: 7,14 g/cm³ (20 °C) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00011291 CAS-nummer: 7440-66-6 EINECS: 231-175-3 Merck Index: 12,10255 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...