Zink ≥99.95% (metals basis), wire, Ø 3.18 mm (0.125 in)
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
10435.G6
10435.H2
10435.G6EA
707
DKK
10435.G6
10435.H2
Zink ≥99.95% (metals basis), wire, Ø 3.18 mm (0.125 in)
Zinc
Purity: 99.95%
CAS No.: 7440-66-6
Physical Form: ≈57.3g/m
Odour: Odourless
MDL No.: MFCD00011291
Formel:
Zn Molekylvægt: 65.39 g/mol Kogepunkt: 908 Smeltepunkt: 420 Massefylde: 7.14 g/cm³ (20 °C) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00011291 CAS-nummer: 7440-66-6 EINECS: 231-175-3 Merck Index: 12,10255 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...