Kobber ≥99.9999% (metals basis), foil, Puratronic®, Thickness: 0.05 mm (0.002 in)

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

42972.FI 42972.GH
42972.FIEA 1540 DKK
42972.FI 42972.GH
Kobber ≥99.9999% (metals basis), foil, Puratronic®, Thickness: 0.05 mm (0.002 in)
Kobber

Purity: 99.9999% CAS No.: 7440-50-8 Physical Form: ≈1.1g/50x50mm Odour: Odourless MDL No.: MFCD00010965

Formel: Cu
Molekylvægt: 63.55 g/mol
Kogepunkt: 2595
Smeltepunkt: 1083
Massefylde: 8.94 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00010965
CAS-nummer: 7440-50-8
EINECS: 231-159-6
Merck Index: 13,02545

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR