Praseodym 99% (REO, rare earth oxide basis), ingot

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

00267.09 00267.18
00267.09EA 1450 DKK
00267.09 00267.18
Praseodym 99% (REO, rare earth oxide basis), ingot
Praseodymium

Purity: 99.9% CAS No.: 7440-10-0 Physical Form: Packaged under mineral oil Sensitivity: Air and Moisture Sensitive Odour: Odourless UN No.: 3178 MDL No.: MFCD00011174

Formel: Pr
Molekylvægt: 140.91 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00011174
CAS-nummer: 7440-10-0
EINECS: 231-120-3
UN nummer: 3178
ADR: 4.1 II

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR