Bly ≥99.999% (metals basis), slug, Puratronic®, Length: 6.35 mm (0.25 in), Ø 6.35 mm (0.25 in)

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

43415.22 43415.14
43415.22EA 6560 DKK
43415.22 43415.14
Bly ≥99.999% (metals basis), slug, Puratronic®, Length: 6.35 mm (0.25 in), Ø 6.35 mm (0.25 in)
Bly

Purity: 99.999% CAS No.: 7439-92-1 Physical Form: ≈2.3g/slug Appearance: Grey Odour: Odourless MDL No.: MFCD00134050

Formel: Pb
Molekylvægt: 207.2 g/mol
Kogepunkt: 1740
Smeltepunkt: 327
Massefylde: 11.3437 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00134050
CAS-nummer: 7439-92-1
EINECS: 231-100-4
Merck Index: 12,05410

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR