Flasker, bred åbning, med skruelåg
Leverandør: Brand
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
128754 128748 128764 128762
BRND128754EA 2100 DKK
BRND128754 BRND128748 BRND128764 BRND128762
Flasker, bred åbning, med skruelåg
Flasker Plastflasker
PFA, translucent, with PFA screw cap.
  • Maximum chemical resistance
  • Heat resistant from –200 to +260 °C
  • Easy to clean
High-quality bottles, specially designed for long-term storage of solvents and standards (trace analysis). PFA screw cap with integrated seal lip and buttress thread.