Amine and carboxyl microplates, Corning® PureCoat™
Leverandør: Corning
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
356717 354719 354717 356719
734-1476EA 5670 DKK
734-1476 734-1477 734-1475 734-1478
Amine and carboxyl microplates, Corning® PureCoat™
Mikroplader Celledyrkningsplader
Corning PureCoat Amine (positively charged) and Carboxyl (negatively charged) surfaces provide improved cell attachment, faster cell proliferation, and enhanced recovery post-thaw over standard TC surfaces. These surfaces function with a broad range of primary, transfected, transformed, and fastidious cell types, and have demonstrated utility in serum-reduced or serum-free conditions.