Pulvertragte med kort, bred stilk
Leverandør: Brand
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
146506 146508 146511 146516 146519 146523
BRND146506EA 107 DKK
BRND146506 BRND146508 BRND146511 BRND146516 BRND146519 BRND146523
Pulvertragte med kort, bred stilk
Tragte Pulvertragte
Borosilicate glass 3,3.
  • Chemically resistant glass
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
  • Quick and easy to clean
Ideal for transferring powders, can also be used for liquids.