Tragte med kort stilk, ribbet
Leverandør: Brand
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
145611 145616 145619 145631 145638
BRND145611EA 4850 DKK
BRND145611 BRND145616 BRND145619 BRND145631 BRND145638
Tragte med kort stilk, ribbet
Tragte Filtreringstragte Filtertragte
Borosilicate glass, ribbed walls.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
For rapid filtration. Cone angle 60°. Funnel lip is ground and fire-polished.