Centrifugerør, EZ Flip™, Nunc™
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
362694 362696 362694NS 362695NS 362696NS 362697NS
734-0995EA 0 DKK
734-0995 734-0997 NUNC362694NS NUNC362695NS NUNC362696NS NUNC362697NS
Centrifugerør, EZ Flip™, Nunc™
Rør Centrifugerør
PP, conical, printed graduations with hinged cap.
  • Printed graduations, white writing area and flat caps for sample identification
  • Hinged-cap system with ergonomic design for one handed opening and closing
  • Integrated leakproof cap prevents cross-contamination and lost closures
  • Withstands forces up to 8500 ×g for 15 ml and 9500 ×g for 50 ml
These tubes are manufactured from high grade polypropylene. RNase-/DNase-free and non pyrogenic.
Emballering: Bagged tubes are packed in durable and re-sealed (zip closure) bags.