Mikroplader, standardhøjde, 384 ShallowWell, Nunc™
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
267461 267462 267460
NUNC267461EA 11310 DKK
NUNC267461 NUNC267462 NUNC267460
Mikroplader, standardhøjde, 384 ShallowWell, Nunc™
Mikroplader HTS mikroplader
PP, non treated. Polypropylene is ideal for storage plates because it has a lower binding capacity to prevent proteins or DNA binding, and allows for complete sample recovery.
  • PP able to withstand temperatures from −80 to +121 °C
  • Rounded wells are optimised for mixing and sample recovery
  • Square rounded wells minimise wicking (capillary action)
  • Natural plates for storage identification or storing colorimetric assays
  • White plates for luminescent assays
  • Black plates for fluorescent assays