Firkantede skåle, 1, 4 og 8 rum, uden gevind, Nunc™
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
267060 267061 267062
734-3301EA 6630 DKK
734-3301 734-3302 734-3303
Firkantede skåle, 1, 4 og 8 rum, uden gevind, Nunc™
Dyrkningsplader Celledyrkningsskåle
Thermo Scientific™ Nunc™ rectangular dishes have standard dimensions and are optimised for robotics.
The one well dish offers a convenient plate format for cell culture and is ideal for HTS Petri dish applications. Four and eight well dishes are ideal for assay compartmentalisation, probe multiple gene subsets and for washing Western blot strips. The four well dish can hold four glass slides for storage. These plates are useful in all areas of cell culture, including scale up and cloning.