HTS 96-multibrønds indsatssystem, Corning® FluoroBlok™
Leverandør: Corning
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
351161 351163 351162 351164
734-0950EA 4410 DKK
734-0950 734-0952 734-0951 734-0953
HTS 96-multibrønds indsatssystem, Corning® FluoroBlok™
Mikroplader Indsatsplader
Corning® FluoroBlok™ HTS 96-multiwell insert system is a cell culture assay platform designed for automation. The one-piece insert housing and fluorescence-blocking microporous membrane (available in 3,0 and 8,0 μm pore sizes) enables increased efficiency, productivity and throughput in the drug discovery process. The receiver plate design minimises crosstalk between wells.
  • Standard technology platform allows multiple protocols
  • Real time detection without dismantling or destroying the insert
  • Eliminates need for manual cell scraping and counting
  • Block >99