VWR® MaximumSF, Støvleovertræk, skridsikker
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
113-1192EA 2500 DKK
113-1192 113-1191 113-1190 113-1189 113-1188 113-1187 113-9107
VWR® MaximumSF, Støvleovertræk, skridsikker
Engangsbeklædning
VWR® Maximum SF boot covers are the cleanest and most durable within the entire VWR® clothing line. Made from spunbonded polypropylene fabric (SPP) and coated with breathable polyethylene film, these garments have been designed to optimise barrier protection, cleanliness and comfort properties.
  • Non skid
  • Impervious material
  • Superior friction coefficient to ensure maximum traction
  • Maximum skid resistance on wet and smooth surfaces
  • Clean, low linting fabric
Testet og fremstillet i ISO-certificerede omgivelser under streng proceskontrol. Lot-kontrolleret og valideret af et uafhængigt testlaboratorium.
Længde: 286 mm (M), 356 mm (U*), 381 mm (XL)
* En størrelse passer alle.