VWR®, Advanced SF Cleanroom Sleeve Protectors (bulk)
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
113-1193EA 1420 DKK
113-1193 113-1194 113-9995 113-9996
VWR®, Advanced SF Cleanroom Sleeve Protectors (bulk)
Engangsbeklædning
VWR® Advanced SF sleeve protectors are manufactured from spunbonded polypropylene fabric (SPP), coated with breathable polyethylene film.
  • Elasticated cuffs, with sonically-welded seams
  • Durable fabric with good breathability and water transmission rate
  • Significant fluid and particle barrier
  • Particle shedding: Level II Helmke drum classification
  • Soft, cloth-like fabric
  • Thumb loop versions are also available
Testet og fremstillet i ISO-certificerede omgivelser under streng proceskontrol. Lot-kontrolleret og valideret af et uafhængigt testlaboratorium.