VWR® Advanced, Kitler, SPP
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
113-1150EA 1010 DKK
113-1150 113-1149 113-1147 113-1146 113-1145 113-1148
VWR® Advanced, Kitler, SPP
Engangsbeklædning
VWR® Advanced SF lab coats are manufactured from spunbonded polypropylene fabric coated with breathable polyethylene film.
  • Tapered collar, inset sleeves, snap closure, elasticated cuffs and three pockets
  • Durable fabric with good breathability and water transmission rate
  • Significant fluid and particle barrier
  • Particle shedding: Level II Helmke drum classification
  • Soft, cloth-like fabric
Testet og fremstillet i ISO-certificerede omgivelser under streng proceskontrol. Lot-kontrolleret og valideret af et uafhængigt testlaboratorium.