Kogebade, SBB Aqua Plus Range
Leverandør: GRANT INSTRUMENTS
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
SBBAQUA12PLUS SBBAQUA26PLUS SBBAQUA5PLUS
462-0166EA 12010 DKK
462-0166 462-0170 462-0171
Kogebade, SBB Aqua Plus Range
Bade Kogebade
Kogevandbade er designet til kontinuerlig drift ved 100 °C. De har en rustfrit stål tank, og analog kontrol ved hjælp af en enkel drejeknap til temperaturindstillinger. Badene har to faste overtemperatur sikkerheds cut-offs der kan nulstilles af brugeren.
  • Energy regulator can be adjusted to ensure a steady boiling action, avoiding excessive steam or spitting
  • Constant level device maintains the required liquid level, avoiding the need to monitor and top up the bath
  • Heater and temperature sensors are mounted at the base of the tank, protected by a perforated tray, giving more usable space
  • Robust construction, durable and resistant to corrosion
  • New modern design
Required accessories must be ordered separately, see SUB Aqua Plus baths entry.
Bestillingsoplysninger: Ønsket tilbehør bestilles seperat.
Leveringsinformation: Leveres uden låg og ringe. Låg anbefales ved temperaturer over 60 °C.
Advarsel: ** Required if flat-bottomed flasks are to be placed directly onto the base of the bath and to allow better convection in the bath.