Vandbad med magnetomrører, HBR 4 control
Leverandør: IKA
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
20003549 10001849
462-0592EA 38340 DKK
462-0592 IKAA20009381 IKAA20009382 IKAA10001849
Vandbad med magnetomrører, HBR 4 control
Bade Vandbade, omrøring
Cylindrical stainless steel bath with heating elements underneath bath vessel. Low viscosity oil 50 cP (mPa.s) or water can be used as the heat transfer fluid. Bath has stirring speed range of 150 to 800 min⁻¹ and the internal tank dimensions are 200×160 mm (Ø×H).
  • Infinitely adjustable safety temperature limiter, according to DIN 12877
  • Double jacket and carrying handles provide protection against accidental burns
  • Fuzzy logic and digital display showing speed and rated, actual and safety temperatures
  • Integrated magnetic stirrer that circulates fluid ensuring improving heat dispersal
  • Safety elements are checked when the unit is switched on
IP protection class according to DIN EN 60529: IP 21
Certificeringer: IP-beskyttelsesklasse i henhold til DIN EN 60529: IP 20
Bestillingsoplysninger: Leveres uden låg og ring sæt. bestil venligst det ønskede tilbehør seperat.
Advarsel: * RT = Rumtemperatur