Olier til vakuumpumper
Leverandør: VACUUBRAND
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
20687010 20687011 20687012 20687013
181-1241EA 401 DKK
181-1241 181-1242 181-1243 181-0449
Olier til vakuumpumper
Smøremidler
Rotary vane pump oil, B
Rotary vane pump oil, B has excellent viscosity characteristics and low vapour pressure. Its exceptional chemical resistance, more stable behaviour when pumping oxidants, acid and alkaline vapours, and the fact that it is better tolerated on contact with skin, sets this oil apart from conventional mineral oils. Even after continuous operation for several weeks and months, the oil ensures the pressure is maintained constantly. Used for priming all RE/RZ series pumps.
Olie K 8
Olie K 8 er en specialolie, der er beregnet specielt til ekstraktion af syreholdige dampe.More Product Information