Vakuum-håndpumpe, Nalgene®
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
6131-0010 6131-0020 6132-0010 6132-0020 6133-0010 6132-2000 6132-3000
181-0434EA 1630 DKK
181-0434 181-9907 181-9902 181-9903 181-9904 181-0473 181-0474
Vakuum-håndpumpe, Nalgene®
Pumper Vakuumhåndpumper
With these sturdy pumps, a partial vacuum of 86 mbar can be reached very quickly and easily. The vacuum can be controlled with just the index finger. Vacuum is set via a lever on the pump. The pumps reach full pressure with just two priming strokes. Removable cover on exhaust port and trigger to release vacuum, so the pump does not need to be disconnected from the partial vacuum line for venting. Hose connection for standard hoses with 6,4 mm internal diameter. Complete with 60 cm transparent plastic tube.
Vakuum-håndpumper uden vakuummåler - PVC-kabinet.
Sugevolumen 36 ml pr. pumpning og 480 mbar overtryk ved udgangsventilen
Økonomisk design, ideel til udendørs brug og i laboratoriet
Inkluderer ikke en vakuummåler
Vakuum-håndpumper med vakuummåler - PVC-kabinet
Sugevolumen 15 ml pr. pumpning og 210 mbar overtryk ved udgangsventilen, art. nr. 181 9902; sugevolumen 36 ml pr. pumpning og 480 mbar overtryk ved udgangsventilen, art. nr. 181 9903
Kan adskilles ved reparation eller udskiftning af indre dele
Inkluderer vakuummåler
Vacuum hand pumps with vacuum gauge, repairable - Zinc alloy housing
Suction volume 15 ml per stroke and 210 mbar overpressure at the outlet valve
Can be disassembled for repair, or to replace internal parts
Includes vacuum gauge