Transilluminators and converter plates, Visi-Blue™
Leverandør: UVP ULTRA VIOLET PRODUCTS
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
38-0200-01 38-0200-02 38-0200-04 95-0433-02 95-0461-02 38-0242-01
730-1019EA 8990 DKK
730-1019 730-1020 UVPA38-0200-04 732-4387 732-4393 UVPA38-0242-01
Transilluminators and converter plates, Visi-Blue™
Lysbord Blåt lys Transilluminatorer
Visi-Blue™ transilluminators are designed for use with SYBR® Gold, SYBR® Green, GFP and other 460 to 470 nm blue excitation samples.
  • Minimise photonicking of DNA samples
  • Amber protective cover blocks blue light transmission and allows visualisation of stained samples above 500 nm
Unit W×D×H: 337×241×121 mm (VB-26V), 486×337×143 mm (VB-40V) (height including blocking cover)
Weight: 6,8 kg (VB-26 V), 9,6 kg (VB-40 V)
Visi-Blue™ converter plates: The Visi-Blue™ converter plate is simply placed on top of a transilluminator equipped with mid-range (3More Product Information