Digital thermometers, KM 330 and KM 340
Leverandør: COMARK
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
KM330 KM340
620-3161EA 1610 DKK
620-3161 620-1174
Digital thermometers, KM 330 and KM 340
Termometre Digital termometre
These multi-purpose thermometers are delivered in a protective rubber boot.
  • Last measurement storage
  • Maximum temperature memory
  • Type KM 340 with twin inputs for direct or differential measurements between two thermocouples
Type K thermocouple probes.