Omrøreraksler og -blade
Leverandør: COWIE
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
002.052.1 002.076.1 002.090.1
441-9561EA 39.5 DKK
441-9561 441-9562 441-9563
Omrøreraksler og -blade
Omrørere Omrøreblade
PTFE. Blades fit PTFE stirring shaft; hole Ø 6,5 mm, 3,2 mm thick. Inert and non-scratch. The smooth surface prevents contamination and makes cleaning easier.
  • Excellent chemical resistance
  • Temperature resistant; suitable for use at high and low temperatures (−200 to +280 °C)