KPG stirrer shafts and stirrer bearings
Leverandør: witeg Labortechnik
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
24.750.09 1.480.000 1.480.016 24.500.42 1.452.000 24.506.57 24.528.56 24.540.51
441-9021EA 1020 DKK
441-9021 441-9101 441-9102 441-9111 441-9112 441-9113 441-9114 441-9115
KPG stirrer shafts and stirrer bearings
Omrørere Omrøreaksler
DURAN®, borosilicate glass interchangeable stirrer shafts and bearings.
  • WB type stirring shafts with ground and polished bearing surface
  • HA, HB, HQ, HT type stirring bearings with ground and polished surface