Magnetic stirrer, remote control, Variomag Mobil 10 and 25
Leverandør: THERMO H + P LABORTECHNIK
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
50119121 50119120
HPLT50119121EA 3640 DKK
HPLT50119121 HPLT50119120
Magnetic stirrer, remote control, Variomag Mobil 10 and 25
Omrørere Magnetomrørere
Disc motor drive stirrers and accessory Telemodul controller, which can stir very large liquid volumes and mix highly viscous media. Stirrers have ultra-flat, space-saving design with housing that is easy-to-clean under running water.
  • Remote control unit allows changes to settings to be made easily, even if stirrer is situated in hazardous or inaccessible areas
  • Maintenance-free drive; automatic start-up for safe stirrer acceleration; quick stop function stops stirring in two seconds to prevent vessels breaking
  • Operating conditions for Mobil 10 are −10 to +56 °C, and −10 to +121 °C for Mobil 25
  • Mobil 25 can be used in water baths, incubators and climatic chambers
  • Telemodul 10 M controller has speed control accuracy of ±1%
IP protection class according to DIN EN 60529: IP 66 (Mobil 10), IP 68 (Mobil 25)
Certificeringer: IP protection class according to DIN EN 60529: IP 66 (Mobil 10), IP 68 (Mobil 25)
Bestillingsoplysninger: Supplied without Telemodul 10 M controller. Please order controller separately.