Manuelt betjent rulle, ledende
Leverandør: BURKLE
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
5309-0010 5309-0020 5309-0030
300-1023EA 1910 DKK
300-1023 300-0015 300-0056
Manuelt betjent rulle, ledende
Håndhjul
The manually operated drum reels are used to lower dipping bottles and immersion cylinders safely to the required depth in shafts, wells and tanks and then to pull them up again. The manually operated drum reels can be equipped with various cables or chains. If flammable or highly flammable liquids are to be collected, then electrically conductive manually operated drum reels must be used together with an earthing cable. The dipping bottles must be made of non sparking metal.
  • High winding capacity of up 150 m for cable 2 mm Ø
  • Reel width: Inner 44 mm, outer 55 mm, reel Ø: Inner 100 mm, outer 180 mm
Bestillingsoplysninger: Please order the required lowering cable separately.