Kemisk øse
Leverandør: BURKLE
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
5353-0250 5353-0500 5353-1000
331-2110EA 1390 DKK
331-2110 331-2111 331-2112
Kemisk øse
Prøvetagere Væskeprøvetagere
Liquid samplers for aggressive liquids, with fixed rod.
  • Three sizes
  • Entirely made of polypropylene − No metal parts
  • Parts coming into contact with sample media made of PP