Prøvetager, MiniSampler
Leverandør: BURKLE
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
5305-0100 5305-0110
300-1021EA 4390 DKK
300-1021 312-1171
Prøvetager, MiniSampler
Prøvetagere Væskeprøvetagere
The sample liquid only comes into contact with the suction hose and the original sample bottle. Cross-contamination is greatly reduced. Immediately after taking the sample, the bottles are sealed.
● PE MiniSampler for general sampling
● PTFE MiniSampler for ultra-pure sampling
  • Suction height depending on the medium (max. 5 m)
  • Disposable suction hose can quickly be replaced to guarantee that the samples are never contaminated
  • Hose can reach narrow seemingly inaccessible sampling areas
MiniSampler PE is suitable for general sampling from barrels, canisters, tanks. Complete ten PE bottles (100 ml), 10 m PE suction hose (6×8 mm).
MiniSampler PTFE is recommended when maximum purity is needed. Complete with a PFA sample bottle (180 ml), 5 m PTFE suction hose (6×8 mm).
Also suitable for viscous oils
Leveringsinformation: In a handy transport case with vacuum pump, hose cutter, stainless steel hose weight made of AISI 304 (1.4301), and 20 seal-it bottle seals.