Lift pumps for chemicals
Leverandør: NRG DEVELOPMENTS
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
5702 5704CHEM
181-0301EA 969 DKK
181-0301 181-0302
Lift pumps for chemicals
Pumper Tromlepumper
These lift pumps feature plastic sleeved steel piston rods so that only compatible materials comes in to contact with the fluid being handled. The standard adapter will fit for 2" BSB or 2" NPT threads. PP, with PTFE seals.
  • Fitted with PTFE seals to offer maximum chemical resistance
  • There are models to fit from 20 to 210 litre containers