Lift pumps for solvents
Leverandør: NRG DEVELOPMENTS
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
5802 5804
181-0303EA 862 DKK
181-0303 181-0304
Lift pumps for solvents
Pumper Tromlepumper
These lift pumps feature plastic sleeved steel piston rods so that only compatible materials comes in to contact with the fluid being handled. The standard adapter will fit for 2" BSB or 2" NPT threads. Nylon, with PTFE seals.
  • Fitted with PTFE seals to offer maximum chemical resistance
  • There are models to fit from 20 to 210 litre containers