Pumper til omhældning, hånd, OTAL®
Leverandør: BURKLE
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
5005-1200 5005-1800 5005-3000 5005-4000 5005-6000 5005-1000 5005-1500 5005-7000
223-0006EA 1550 DKK
223-0006 223-1112 223-1115 223-0061 223-0060 223-1130 223-1113 223-1000
Pumper til omhældning, hånd, OTAL®
Pumper Tromlepumper
The telescopic immersion tube adapts itself to the container height on insertion. This ensures good removal of residual liquid and means that the hand pump can be used with different containers.
  • Particularly compact, space-saving hand pump
  • Good pump capacity
  • With universal bungs made of soft PVC, for container openings of 30 to 70 mm Ø
OTAL® hand pump made of PP
OTAL® hand pumps with PP delivery tubes, resistant to weak acids, alkalis and detergents.
OTAL® hand pump made of PVDF
OTAL® hand pump with PVDF delivery tubes, particularly suitable for acids and alkalis, not for heavily oxidising acids and alkalis. With its special material properties, PVDF has an even better chemical resistance than PP. Resistant to chlorine bleach.
OTAL® hand pump made of stainless steel
OTAL® hand pump with delivery tubes made of stainless steel AISI 304 (1.4301) for liquids that are not easily combustible in Hazard Category AIII (VbF). The OTAL® PP or PVDF hand pump should be used for caustic media.
OTAL® hand pump with hose and stopcock
OTAL® hand pump with hose and tap with discharge hose with a length of 1,5 m made of PVC and PP/PE stopcock, delivery tube made of PP. The discharge hose is particularly suited to fill distant containers.
OTAL® mini pump made of PP
OTAL® PP mini pump with delivery tubes made of PP and universal bungs made of soft PVC. Suitable for small containers up to 10 litres. With universal bungs for container openings of 20 to 36 mm Ø
OTAL® battery-pump
OTAL® pump for safe filling of car batteries with acid or distilled water, with hose shutoff valve and dosing tip for precise filling.