Digitale polarimetre, AP-300
Leverandør: Atago
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
5291 RE-72043 RE-72044 RE-72045
635-0409EA 58790 DKK
635-0409 635-0500 635-0501 635-0502
Digitale polarimetre, AP-300
Polarimetre
AP-300 with automatic temperature compensation (ATC) for sugar factories.
  • Password function to identify operators and protect data
  • Built-in International Sugar Scale (ISS)
  • Manual calibration with quartz control plates
Temperature control is not necessary for users who have temperature controlled working environments or are able to benefit from the temperature compensation funtionality.
The Pharmaceutical packages include a GMP and GLP compliant quartz control plate for calibration and a digital printer DP-AD (regular paper) for data recording.
Certificeringer: ICUMSA compliant - FDA 21 CFR Part 11 software.
Leveringsinformation: Leveres med 100 mm og 200 mm observationsrør