Enkeltkanalpipetter, elektroniske, Transferpette® electronic
Leverandør: Brand
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
705300 705303 705306 705340 705343 705346 705316 705313 705310 705307 705299 705339 705347 705309 705317 705319 705320 705323 705326 705330
613-4217EA 4500 DKK
613-4217 613-4218 613-4219 613-4223 613-4224 613-4225 613-4222 613-4221 613-4220 613-0083 613-0081 613-0195 613-0196 613-0596 613-0597 BRND705319 BRND705320 BRND705323 BRND705326 BRND705330
Enkeltkanalpipetter, elektroniske, Transferpette® electronic
Pipetter Automatiske pipetter
The piston-operated pipette Transferpette® electronic combines the widely recognized features of BRAND mechanical pipettes with the advantages of electronic apparatus.
  • Easy to use: Intuitive menu control, coloured eject button and straightforward technical documentation
  • Due to the optimised standard design of the tip cone, BRAND® or other manufacturers' tips can be used
  • Wide selection of programs; pipetting, sample mixing, reverse pipetting, pipetting for electrophoresis, dispensing, ‘Easy Calibration' program for making quick adjustments to the Transferpette® electronic without tools, and ‘Refresh' battery functions
  • Autoclavable (pipette shaft only)
* Filter
Bestillingsoplysninger: Supplied with certificate of quality, battery, and silicone oil.