VWR®, Enkeltkanalpipetter, mekaniske, fast volumen
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
613-0172EA 1440 DKK
613-0172 613-7119 613-7120 613-7121 613-7122 613-7123 613-7124 613-7125 613-7126 613-7127 613-7128
VWR®, Enkeltkanalpipetter, mekaniske, fast volumen
Pipetter Automatiske pipetter
Ergonomically designed to fit large or small hands for extra comfort while working, for left or right handed operation.
  • Slim tip mounting cone compatible with all tips
  • Stainless steel movable and height-adjustable tip ejector
  • Maintenance-free seals, polished plungers for superior precision
  • Fully autoclavable
Bestillingsoplysninger: Leveres med en nøgle til kalibrering i laboratoriet.