Accessories for Ultra-centrifugal mill, ZM 200
Leverandør: Retsch GmbH
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
22.524.0001 02.186.0010 02.010.0037 02.010.0039 22.786.0002 03.010.0041 22.355.0005 22.261.0003 03.010.0037 05.708.0065 02.011.0031 22.355.0006 22.355.0003 22.935.0015 22.935.0016 22.935.0019 22.935.0014 22.935.0017 22.936.1001 22.355.0009 02.011.0029 05.742.0017 03.111.0201
RETS22.524.0001EA 426 DKK
RETS22.524.0001 412-2133 RETS02.010.0037 412-0010 412-2933 412-2937 RETS22.355.0005 412-2934 RETS03.010.0037 RETS05.708.0065 RETS02.011.0031 RETS22.355.0006 RETS22.355.0003 412-0076 412-0233 RETS22.935.0019 RETS22.935.0014 RETS22.935.0017 412-1006 RETS22.355.0009 RETS02.011.0029 RETS05.742.0017 RETS03.111.0201
Accessories for Ultra-centrifugal mill, ZM 200
Møller
The ZM 200 is a powerful ultra-centrifugal mill for both demanding grinding tasks and efficient routine work. ZM 200 reduces the size of soft to medium-hard and fibrous materials. Ideal for rapid yet gentle preparation of analytical samples of materials such as chemicals, drugs, spices, coal, synthetic resins, plastics, pharmaceutical raw materials and finished products, plant constituents and surfactant determination in washing powders. For grinding larger amounts the use of a paper filter bag or a cyclone with 3 or 5 litre collecting receptacle is recommended. The sample is then cooled by the airstream and discharged more rapidly from the grinding chamber via the passage receptacle.
  • Ground material is collected in the cassette, which ensures easy and loss-free material removal and avoids cross contamination
  • Versatile mill with wide speed range; speed can be individually selected from 6000 to 18 000 min-1
  • Increased throughput due to optimally matched frequency converter and three phase motor
  • Two step grinding ensures gentle but fast processing without altering sample characteristics
  • Parameters are set via a graphics display and one button operation, electronic safety and diagnosis system reduce operating errors
IP protection class according to DIN EN 60529: IP 20
* Depending on feed material